EMAIL THIS PAGE TO A FRIEND
Oncotarget
Notch3 overexpression enhances progression and chemoresistance of urothelial carcinoma.
PMID 28416766
Abstract
Abnormal activation of Notch signaling is involved in the etiology of various diseases, including cancer, but the association between Notch3 expression in urothelial cancer and clinical outcome remains unclear, and the molecular mechanisms underlying Notch3 signaling activation are not well defined. In this study we examined 59 urothelial cancer patients and found that Notch3 was more highly expressed in human urothelial cancer tissues than in non-tumorous bladder tissue samples, with Notch3 overexpression being associated with poor clinical outcome. Notch3 knockdown resulted in decreased proliferation of urothelial cancer cells in vitro and decreased xenograft tumor growth in vivo. In addition, Notch3 knockdown rendered urothelial cancer cells more sensitive to cisplatin. Furthermore, suberoylanilide hydroxamic acid (SAHA, a histone deacetylase [HDAC] inhibitor) induced acetylation of NOTCH3, downregulated Notch 3, prevented urothelial cancer cell proliferation, and induced cell cycle arrest. Taken together, these data suggested that Notch 3 overexpression promotes growth and chemoresistance in urothelial cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0061
SAHA, ≥98% (HPLC)
C14H20N2O3
C2211
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
C26H41N3O5