EMAIL THIS PAGE TO A FRIEND
Journal of food science and technology
Supercritical CO
PMID 28416879
Abstract
A series of experiments was conducted to determine optimum conditions for supercritical carbon dioxide extraction of candlenut oil. A Taguchi experimental design with L
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7517
Glyceryl tridecanoate, ≥99% (GC)
C33H62O6