EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
Exploring the biological activity of condensed tannins and nutritional value of tree and shrub leaves from native species of the Argentinean Dry Chaco.
PMID 28417450
Abstract
Tropical tree or shrub leaves are an important source of nutrients for ruminants and a potential source of biologically active compounds that may affect ruminal metabolism of nutrients. Therefore, eight woody species from the native flora of Argentinean Dry Chaco, rich in secondary compounds such as condensed tannins (CT), were assessed for their nutritional value, CT fractions and in vitro true digestibility of dry matter, as well as biological activity (BA). Differences among species were found in contents of total phenol, protein-precipitating phenols (PPP), bound proteins to PPP (BP) and BP/PPP (P < 0.0001). The BP/PPP ratio reveals differences among species in potential BA as indicated by protein precipitation. The major CT of each species were isolated and purified for use as a standard. Although Schinopsis balansae had the most (P ≤ 0.05) total CT (19.59% DM), Caesalpinia paraguariensis had greater (P ≤ 0.05) BA with the most PPP (530.21% dry matter). Larrea divaricata, at 0.97, followed by Acacia aroma, at 0.89, had CT with the highest (P ≤ 0.05) BP/PPP ratios, followed by Prosopis alba (0.59). There were differences in nutritive value and bioactivity among species. Those with the greatest CT were not necessarily those with the most BA. Caesalpinia paraguariensis, S. balansae and L. divaricata were the most promising species as native forage CT sources. Cercidiurm praecox (20.87% CP; 18.14% acid detergent fiber) and Prosopis nigra (19.00% CP; 27.96% acid detergent fiber) showed the best (P ≤ 0.05) nutritive values. According to their nutritive traits, these species might be complementary in grass-based ruminant diets. © 2017 Society of Chemical Industry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1659
Pelargonidin chloride
C15H11ClO5