EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
An efficient process for obtaining prebiotic oligosaccharides derived from lactulose using isomerized and purified whey permeate.
PMID 28417455
Abstract
One of the most promising uses of whey permeate (WP) is the synthesis of prebiotic oligosaccharides. Herein, commercial WP was submitted to chemical isomerization catalysed by sodium borate at an alkaline pH and subsequent purification using anion-exchange resins to remove boron. Subsequently, purified mixtures were used to synthesize prebiotic oligosaccharides using β-galactosidase from Bacillus circulans. Isomerization of concentrated WP (200 g L This study shows that combined chemical-enzymatic reactions together with the purification of IWP results in an efficient synthesis of prebiotic oligosaccharides. © 2017 Society of Chemical Industry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92771
3-(Trimethylsilyl)-2-oxazolidinone, ≥97.0%
C6H13NO2Si