EMAIL THIS PAGE TO A FRIEND
Journal of basic microbiology
Recombinant overexpression of dihydroneopterin aldolase catalyst potentially regulates folate-biofortification.
PMID 28418068
Abstract
We aim to investigate the prospects of increased production of folate through the overexpression of heterologous dihydroneopterin aldolase catalyst. The gene encoding aldolase catalyst was cloned into an expression vector and the induced recombinant protein was purified through metal-affinity chromatography which appeared at 14 kDa position on polyacrylamide-gel. Remarkably, a periodic increase in the extracellular and intracellular folic acid concentration was observed at 4 h growth of induced recombinant DHNA samples than control in a pH-dependent manner. Maximum folate concentration was observed with at least twofold increase in induced recombinant samples at pH8.0 compared to the significant decline at 6 h growth. Consistently, heterologous overexpression of bacterial aldolase through Agrobacterium-mediated genetic transformation of tobacco led to more than 2.5-fold increase in the folate concentration in the transgenic leaves than control tissues. These data are veritable inspecting metabolic flux in both bacterial and plant systems, thus providing directions for future research on folate agri-fortification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1278
Galantide, ≥90% (HPLC)
C104H151N25O26S