EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Modular Flow-Through Platform for Spectroelectrochemical Analysis.
PMID 28418648
Abstract
A new type of flow platform for electrochemical and spectroelectrochemical measurements is presented. Finite element method simulations confirm that the hydrodynamic profile within the device is not turbulent and provides an analytical platform for the investigation of homogeneous kinetics, radical lifetimes, and reaction mechanisms. The modular "plug and play" configuration of the platform allows one to carry out electrochemistry and spectroscopy individually or simultaneously. Specific demonstrations of electroanalytical measurements using the flow system platform includes voltammetric analysis of organometallic compounds and quantitative analysis of ascorbic acid in commercial orange juice samples. Combined spectroelectrochemical demonstrations include electrochemical luminescence of ruthenium compounds and ligand exchange reactions of iron complexes using UV-vis spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
335061
Ferrocenemethanol, 97%
C11H12FeO