EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
The Heme Metabolite Carbon Monoxide Facilitates KSHV Infection by Inhibiting TLR4 Signaling in Endothelial Cells.
PMID 28421060
Abstract
Kaposi sarcoma herpesvirus (KSHV) is the etiologic agent of Kaposi sarcoma (KS) and certain rare B cell lymphoproliferative disorders. KSHV infection of endothelial cells (EC)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
288144
Tricarbonyldichlororuthenium(II) dimer
C6Cl4O6Ru2