La Radiologia medica
Whole body MRI with qualitative and quantitative analysis of DWI for assessment of bone marrow involvement in lymphoma.
PMID 28421406
Our study aimed to investigate the role of qualitative and quantitative whole body MRI with DWI for assessment of bone marrow involvement (BMI) in newly diagnosed lymphoma using FDG PET-CT and bone marrow biopsy (BMB) as reference standard. We retrospectively evaluated 56 patients with newly diagnosed lymphoma (21 Hodgkin's lymphoma and 35 non-Hodgkin's lymphoma) who underwent random unilateral BMB, FDG PET-CT and Wb-MRI-DWI for initial staging. In a patient-based analysis, results of Wb-MRI-DWI were compared with FDG PET-CT and BMB. For quantitative analysis, mean ADC values of posterior iliac crest were correlated with BMI and bone marrow cellularity. WB-MR-DWI obtained excellent concordance with FDG PET-CT both in HL (k = 1.000; 95% CI 1.000-1.000) and in DLBCL (k = 1.000; 95% CI 1.000-1.000). In other NHL, WB-MRI-DWI obtained a good correlation with BMB (k = 0.611; 95% CI 0.295-0.927) while FDG PET-CT had poor concordance (k = 0.067; 95% CI 0.372-0.505). WB-MR-DWI has no false negative errors but 4 false positive results consisting in focal lesions consensually reported by FDG PET-CT and resolved after therapy. No significant correlation between ADC mean value and BMI was found (p = 0.0586). Our data suggest that Wb-MRI-DWI is a valid technique for BMI assessment in lymphoma patients, thanks to its excellent concordance with FDG PET-CT and good concordance with BMB (superior than FDG PET-CT). If further investigations will confirm our results on larger patient groups, it could become a useful tool in the clinical workup.
Related Materials
Product #
Molecular Formula
Add to Cart
1,4-Di(maleimido)butane, ≥99.0% (HPLC)