EMAIL THIS PAGE TO A FRIEND
Oncotarget
Epigenetic silencing of miR-520c leads to induced S100A4 expression and its mediated colorectal cancer progression.
PMID 28423501
Abstract
The S100 calcium-binding protein A4 (S100A4) induces epithelial mesenchymal transition, migration, invasion, angiogenesis and metastasis. Its induced expression in several cancer types correlates with poor prognosis. Apart from the functional and transcriptional regulatory aspects of S100A4, its post-transcriptional regulation is not yet clearly elucidated. In this study, we show that microRNAs (miR) miR-505-5p and miR-520c-3p target the 3'-UTR of S100A4 and inhibits its expression and its mediated migration and invasion. 5-Aza treatment significantly increased miR-520c-3p expression and reduced the S100A4 protein amounts. The upstream promoter region of miR-520c is hypermethylated irrespective of the metastasis status of colorectal cancer (CRC) patient tissues and in all analyzed CRC cell lines. Moreover, in a cohort of CRC patient specimen (n = 59), miR-520c-3p was significantly downregulated. miR-520c-3p stably expressing HCT116 cells showed a reduced metastasis formation in livers after implanting in mice spleen. Taken together, our findings demonstrate that S100A4 is post-transcriptionally regulated by tumor suppressor miRs, miR-505c-5p and miR-520c-3p, and particularly miR-520c-3p expression is epigenetically silenced in CRC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU125681 MISSION® esiRNA, esiRNA human S100A4 (esiRNA1)