EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
UCP2 attenuates apoptosis of tubular epithelial cells in renal ischemia-reperfusion injury.
PMID 28424210
Abstract
Uncoupling protein-2 (UCP2) plays critical roles in energy metabolism and cell survival. Previous investigations showed that UCP2 regulated the production of extracellular matrix and renal fibrosis. However, little is known about UCP2 in acute kidney injury (AKI). Here, we used