Ultrasonics sonochemistry
Combined effect of ultrasound, heat, and pressure on Escherichia coli O157:H7, polyphenol oxidase activity, and anthocyanins in blueberry (Vaccinium corymbosum) juice.
PMID 28427631
The objective of this study was to evaluate the effect of different treatments-heat treatment (HT), sonication (SC), thermosonication (TS), manosonication (MS), manothermal (MT), and manothermosonication (MTS) on Escherichia coli O157:H7, polyphenol oxidase (PPO), and anthocyanin content in blueberry juice. First, samples were treated at different temperatures (30, 40, 50, 60, 70, and 80°C) and power intensities (280, 420, 560, and 700W) for 10min. Subsequently, samples were treated using combinations of power intensity and mild temperature for 10min. For further study, samples were treated using HT (80°C), TS (40°C, 560W), MT (350MPa, 40°C), MS (560W, 5min/350MPa), or MTS (560W, 5min, 40°C/350MPa, 40°C) for 5, 10, 15, 20min for each treatment, and the results compared between treatments. HT significantly reduced PPO activation (2.05% residual activity after only 5min), and resulted in a 2.00-log reduction in E. coli O157:H7 and an 85.25% retention of anthocyanin. Escherichia coli O157:H7 was slightly inactivated by TS after 5min (0.17-log reduction), while residual PPO activity was 23.36% and anthocyanin retention was 98.48%. However, Escherichia coli O157:H7 was rapidly inactivated by MTS (5.85-log reduction) after 5min, while anthocyanin retention was 97.49% and residual PPO activity dropped to 10.91%. The destruction of E. coli cells as a result of these treatments were confirmed using SEM and TEM. Therefore, a combination of sonication, high pressure, and mild heat allows the safety of blueberry juice to be maintained without compromising the retention of desirable antioxidant compounds.
Related Materials
Product #
Molecular Formula
Add to Cart
Suplatast tosylate, ≥98% (HPLC)
C16H26SNO4 · C7H7SO3