EMAIL THIS PAGE TO A FRIEND
Journal of biotechnology
Enhancement of ethyl (S)-4-chloro-3-hydroxybutanoate production at high substrate concentration by in situ resin adsorption.
PMID 28427921
Abstract
Asymmetric reduction of ethyl 4-chloro-3-oxobutyrate (COBE) by carbonyl reductases presents an efficient way to produce Ethyl (S)-4-chloro-3-hydroxybutanoate ((S)-CHBE), an important chiral intermediate for the synthesis of hydroxymethylglutaryl-CoA reductase inhibitors such as Lipitor