EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Nitric oxide-regulated proteolysis of human CYP2B6 via the ubiquitin-proteasome system.
PMID 28427998
Abstract
We showed previously that rat cytochrome P450 CYP2B1 undergoes NO-dependent proteasomal degradation in response to inflammatory stimuli, and that the related human enzyme CYP2B6 is also down-regulated by NO in primary human hepatocytes. To investigate the mechanism of CYP2B6 down-regulation, we made several cell lines (HeLa and HuH7 cells) in which native CYP2B6 or CYP2B6 with a C-terminal V5 tag (CYP2B6V5) are expressed from a lentiviral vector with a cytomegalovirus promoter. Native CYP2B6 protein was rapidly down-regulated in HeLa cells within 3h of treatment with the NO donor (Z)-1-[2-(2-Aminoethyl)-N-(2-ammonioethyl)amino]diazen-1-ium-1,2-diolate, while its mRNA level was not down-regulated. Treatment of the cells with the NO donor (Z)-1-[N-(3-aminopropyl)-N-(3-ammoniopropyl)amino]diazen-1-ium-1,2-diolate also resulted in rapid down-regulation of CYP2B6 activity, measured as the formation of 7-hydroxy-4-trifluoromethylcoumarin, as well as 2B6 protein in the CYP2B6 HeLa cell line. CYP2B6V5 was also down-regulated by NO donors in HuH7 cells. Down-regulation was observed in the presence of cycloheximide, demonstrating that this occurs via a post-translational mechanism. We generated a HeLa cell line expressing both CYP2B6V5 and human nitric oxide synthase 2 (NOS2), the latter under positive control by tetracycline. The cellular NO produced by doxycycline treatment also effectively down-regulated CYP2B6 protein, which was blocked by the co-treatment with the NOS2 competitive inhibitor L-N