EMAIL THIS PAGE TO A FRIEND
Molecular biology of the cell
RABL2 interacts with the intraflagellar transport-B complex and CEP19 and participates in ciliary assembly.
PMID 28428259
Abstract
Proteins localized to the basal body and the centrosome play crucial roles in ciliary assembly and function. Although RABL2 and CEP19 are conserved in ciliated organisms and have been implicated in ciliary/flagellar functions, their roles are poorly understood. Here we show that RABL2 interacts with CEP19 and is recruited to the mother centriole and basal body in a CEP19-dependent manner and that CEP19 is recruited to the centriole probably via its binding to the centrosomal protein FGFR1OP. Disruption of the