EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Electrocatalytic dechlorination of 2,3,5-trichlorophenol on palladium/carbon nanotubes-nafion film/titanium mesh electrode.
PMID 28429272
Abstract
Palladium/carbon nanotubes-nafion film-modified titanium mesh electrode (Pd/CNTs-nafion film/Ti electrode) was prepared and used for catalytic dechlorination of 2,3,5-trichlorophenol (2,3,5-TCP). The influences of factors, such as Pd
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442895
Mercury(I) sulfate, 96%
Hg2O4S