EMAIL THIS PAGE TO A FRIEND
Reproductive toxicology (Elmsford, N.Y.)
Thyroid disrupting pesticides impair the hypothalamic-pituitary-testicular axis of a wildlife bird, Amandava amandava.
PMID 28431984
Abstract
The effect of two thyroid disrupting pesticides (TDPs) mancozeb (MCZ) and imidacloprid (IMI) on the hypothalamic-pituitary-gonadal/testicular (HPG) axis of a seasonally breeding bird, Amandava amandava has been evaluated. Male birds (n=8/group) were exposed to each of the pesticide (0.25% LD