EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Exploring Non-obvious Hydrophobic Binding Pockets on Protein Surfaces: Increasing Affinities in Peptide-Protein Interactions.
PMID 28432776
Abstract
A 42-residue polypeptide conjugated to a small-molecule organic ligand capable of targeting the phosphorylated side chain of Ser15 was shown to bind glycogen phosphorylase a (GPa) with a K
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47691
Fmoc-α-Me-Ala-OH, ≥97.0% (HPLC)
C19H19NO4
47428
Fmoc-Phe(4-F)-OH, ≥98.0% (HPLC)
C24H20FNO4