EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Design, selection, and development of cyclic peptide ligands for human erythropoietin.
PMID 28433433
Abstract
This work presents the selection and characterization of erythropoietin (EPO)-binding cyclic peptide ligands. The sequences were selected by screening a focused library of cyclic depsipeptides cyclo[(N
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
335150
Phenylsilane, 97%
C6H8Si
233781
Triisopropylsilane, 98%
C9H22Si