EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Effect of 1,2,3-triazole salts, non-classical bioisosteres of miltefosine, on Leishmania amazonensis.
PMID 28433512
Abstract
Here, we report the effect of new non-classical bioisosteres of miltefosine on Leishmania amazonensis. Fifteen compounds were synthesized and the compound dhmtAc, containing an acetate anion, a side chain of 10 carbon atoms linked to N-1 and a methyl group linked to N-3, showed high and selective biological activity against L. amazonensis. On the intracellular amastigotes, stages of the parasite related to human disease, the IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4069
JC-1, solid
C25H27Cl4IN4