EMAIL THIS PAGE TO A FRIEND
Nuclear medicine and biology
Detection of melanoma metastases with PET-Comparison of
PMID 28433794
Abstract
Cervical LN and lung melanoma metastasis models were established in C57BL/6 mice. Primary tumors were created by injection of melanoma cells into the pinna, and the resulting cervical LN metastases were evaluated. Lung metastases were created by intravenous injection of melanoma cells. The mice underwent LN metastases with a diameter<1cm were more visible on
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7791
N-Acetyl-D-lactosamine, ≥98%
C14H25NO11