EMAIL THIS PAGE TO A FRIEND
Acta histochemica
Resveratrol improves mitochondrial function in the remnant kidney from 5/6 nephrectomized rats.
PMID 28434671
Abstract
Mitochondrial dysfunction is involved in the pathogenesis of chronic kidney disease (CKD). Resveratrol has been demonstrated to be beneficial for the recovery of kidney diseases. In this study, the 5/6 nephrectomized rat was used as a CKD model and the TGF-β1-exposed mouse mesangial cells were used as an in vitro model. Pathological examination showed that resveratrol treatment attenuated glomerular injury in the remnant kidney of 5/6 nephrectomized rat. Additionally, resveratrol improved mitochondrial function in vivo and in vitro, as evidenced by increasing mitochondrial membrane potential, increasing ATP, decreasing reactive oxygen species production and enhancing activities of complex I and III. Furthermore, the dysregulated expressions of electron transport chain proteins and fission/fusion proteins in the kidney of 5/6 nephrectomize rats and TGF-β1-exposed mesangial cells were restored by resveratrol. Finally, upregulated sirt1 and PGC-1α deacetylation were found after treatment with resveratrol in vivo and in vitro, which may contribute to the mitochondrial protective effects of resveratrol. The results demonstrate that resveratrol protects the mitochondria of kidney in 5/6 nephrectomized rats and TGF-β1 induced mesangial cells. The study provides new insights into the renoprotective mechanisms of resveratrol.