EMAIL THIS PAGE TO A FRIEND
Fungal genetics and biology : FG & B
The mitogen-activated protein kinase GlSlt2 regulates fungal growth, fruiting body development, cell wall integrity, oxidative stress and ganoderic acid biosynthesis in Ganoderma lucidum.
PMID 28435030
Abstract
The mitogen-activated protein kinases (MAPKs) are crucial signaling instruments in eukaryotes that play key roles in regulating fungal growth, development, and secondary metabolism and in adapting to the environment. In this study, we characterized an Slt2-type MAPK in Ganoderma lucidum, GlSlt2, which was transcriptionally induced during the primordium and fruiting body stages. RNA interference was used to examine the function of GlSlt2. Knockdown of GlSlt2 caused defects in growth and increased hyphal branching as well as hypersensitivity to cell wall-disturbing substances. Consistently, the chitin and β-1,3-d-glucan contents and the expression of cell wall biosynthesis genes were decreased and down-regulated, respectively, in GlSlt2 knockdown strains compared with those in the wild type (WT). In addition, no primordium or fruiting body could be observed in GlSlt2 knockdown strains. Furthermore, the intracellular reactive oxygen species (ROS) content and ganoderic acid biosynthesis also decreased in GlSlt2 knockdown strains. Addition of H