EMAIL THIS PAGE TO A FRIEND
Water research
Distinguishing homogeneous-heterogeneous degradation of norfloxacin in a photochemical Fenton-like system (Fe
PMID 28436822
Abstract
This study demonstrated the efficient degradation of a typical bio-refractory antibiotic norfloxacin (NOR) in a photochemical iron oxides/oxalate system adopting magnetic catalyst (Fe
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131377
1,10-Phenanthroline, ≥99%
C12H8N2
N9890
Norfloxacin, analytical standard, ≥98% (TLC)
C16H18FN3O3