Cellular immunology
Progression of experimental autoimmune encephalomyelitis is associated with up-regulation of major sodium transporters in the mouse kidney cortex under a normal salt diet.
PMID 28438314
Recent demonstrations of exacerbation of experimental autoimmune encephalomyelitis (EAE) by high salt diets prompted us to study whether EAE stimulated Na absorption by the renal cortex, a primary regulatory site for Na balance, even under a normal NaCl diet. We found that as EAE progressed from mild to severe symptoms, there were parallel increases in the protein abundance of NHE3 and αENaC and the Na,K-ATPase activity with an affiliated elevation of its β1-subunit protein. These effects are associated with increases in the protein levels of the well-known regulators SGK1 and scaffold NHERF2, and phosphorylation of ERK1/2. These effects of EAE could not be explained by reduction in water or food intake. We conclude that EAE progression is associated with up-regulation of major Na transporters, which is most likely driven by increased expression of SGK1 and NHERF2 and activation of ERK1/2. These data suggest that EAE progression increases Na absorption by the renal cortex.