Neurotoxicology and teratology
Acute effects of amitriptyline on adult zebrafish: Potential relevance to antidepressant drug screening and modeling human toxidromes.
PMID 28438663
The need to develop novel antidepressants is an emerging problem in biomedicine. An aquatic vertebrate species, the zebrafish (Danio rerio) may serve as a useful in-vivo screen for CNS drugs, and displays high sensitivity to a wide range of antidepressants. Amitriptyline is a commonly used tricyclic antidepressant which acts primarily as a serotonin and noradrenaline reuptake inhibitor. Here, we characterize drug-induced behavioral and neurochemical responses in adult zebrafish following their acute exposure to amitriptyline. Overall, the drug at 1 and 5mg/L significantly increased time spent in top and shortened the latency to enter it, thereby paralleling recent reports on zebrafish 'serotonin toxicity-like behavior' caused by various serotonergic agents. The 10mg/L dose of the drug also significantly decreased top entries and maximal velocity and evoked overt ataxia, likely due to emerging non-specific toxic effects. Amitriptyline at 5 and 10mg/L also dose-dependently increased serotonin turnover, but not noradrenaline levels, in zebrafish whole-brain samples. Overall, zebrafish high sensitivity to acute effects of amitriptyline can help improve our understanding of psychopharmacological profiles of this compound and the related CNS drugs, and contributes further to the development of aquatic experimental models of human toxidromes.