EMAIL THIS PAGE TO A FRIEND
Chirality
Factors screening to statistical experimental design of racemic atenolol kinetic resolution via transesterification reaction in organic solvent using free Pseudomonas fluorescens lipase.
PMID 28439969
Abstract
As the (R)-enantiomer of racemic atenolol has no β-blocking activity and no lack of side effects, switching from the racemate to the (S)-atenolol is more favorable. Transesterification of racemic atenolol using free enzymes investigated as a resource to resolve the racemate via this method is limited. Screenings of enzyme, medium, and acetyl donor were conducted first to give Pseudomonas fluorescens lipase, tetrahydrofuran, and vinyl acetate. A statistical design of the experiment was then developed using Central Composite Design on some operational factors, which resulted in the conversions of 11.70-61.91% and substrate enantiomeric excess (ee) of 7.31-100%. The quadratic models are acceptable with R
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
330884
(R)-(+)-Atenolol, 99%
C14H22N2O3