EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Dawson-Type Polyoxomolybdate Anions (P
PMID 28440567
Abstract
The development of advanced anode materials is of significance for the next generation of lithium-ion batteries (LIBs). Herein, imidazolium-based ionic liquids (ILs) are grafted covalently on the surface of graphene oxides (GO) through the carboxyl-amino bond, providing a loading hotbed for the Dawson-type polyoxometalate (POM) of (NH