EMAIL THIS PAGE TO A FRIEND
Pharmacology, biochemistry, and behavior
Further evaluation of the potential anxiolytic activity of imidazo[1,5-a][1,4]diazepin agents selective for α2/3-containing GABA
PMID 28442369
Abstract
Positive allosteric modulators of GABA