EMAIL THIS PAGE TO A FRIEND
Biomedical research (Tokyo, Japan)
Histochemical assessment for osteoblastic activity coupled with dysfunctional osteoclasts in c-src deficient mice.
PMID 28442663
Abstract
Since osteoblastic activities are believed to be coupled with osteoclasts, we have attempted to histologically verify which of the distinct cellular circumstances, the presence of osteoclasts themselves or bone resorption by osteoclasts, is essential for coupled osteoblastic activity, by examining c-fos