EMAIL THIS PAGE TO A FRIEND
Frontiers in plant science
Transcriptome-Guided Mining of Genes Involved in Crocin Biosynthesis.
PMID 28443112
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17304
Crocin, for microscopy
C44H64O24