EMAIL THIS PAGE TO A FRIEND
The Journal of nutritional biochemistry
Vitamin D supplementation restores the blunted muscle protein synthesis response in deficient old rats through an impact on ectopic fat deposition.
PMID 28445792
Abstract
We investigated the impact of vitamin D deficiency and repletion on muscle anabolism in old rats. Animals were fed a control (1 IU vitamin D