EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Murinization and H Chain Isotype Matching of the Anti-GITR Antibody DTA-1 Reduces Immunogenicity-Mediated Anaphylaxis in C57BL/6 Mice.
PMID 28446565
Abstract
Recent advances in immuno-oncology have shown that the immune system can be activated to induce long-term, durable antitumor responses. For immuno-oncology drug development, immune activation is often explored using rat Abs in immunocompetent mouse models. Although these models can be used to show efficacy, antidrug immune responses to experimental protein-based therapeutics can arise. Immunogenicity of surrogate Abs may therefore represent an important obstacle to the evaluation of the antitumor efficacy of immunomodulator Abs in syngeneic models. A recent publication has shown that anti-glucocorticoid-induced TNFR family-related protein agonistic Ab DTA-1 (rat or murinized IgG2a) can induce the development of anaphylaxis in C57BL/6 mice upon repeated i.p. dosing because of an anti-idiotypic anti-drug Ab immune response. This study was undertaken to address the impact of the immunogenicity derived from the Fc and variable domains. To this end, chimerized (rat V domains/mouse constant regions) and murinized (95% mouse sequence) DTA-1-based surrogate Abs with a murine IgG2c H chain isotype were created. Chimerization and murinization of DTA-1 did not affect receptor binding and glucocorticoid-induced TNFR family-related protein-induced T cell agonistic properties. Similar in vivo antitumor efficacy and intratumoral CD8
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0799 Mouse IgE ELISA Kit