EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Efficiency and Safety of β-CD-(D3)7 as siRNA Carrier for Decreasing Matrix Metalloproteinase-9 Expression and Improving Wound Healing in Diabetic Rats.
PMID 28447455
Abstract
Overexpression of matrix metalloproteinase-9 (MMP-9) is critical for diabetic chronic wounds involved in the refractory wound healing process. We aimed to develop a strategy through RNAi to decrease MMP-9 expression and improve diabetic wound healing. We had explored β-CD-(D3)7 as a gene carrier to take siRNA and effectively interfere with MMP-9 expression. It has been proven that β-CD-(D3)7 could be used as an effective siRNA delivery system. In this study, we want to know about the efficiency and safety of β-CD-(D3)7/MMP-9 siRNA for improving wound healing in diabetic rats. β-CD-(D3)7/MMP-9 siRNA treated animals show lower levels of MMP-9 expression, which induce faster wound-close rates. Histological evaluation indicates that β-CD-(D3)7/MMP-9 siRNA significantly increases the content of collagen around the injured tissues. The number of neutrophilic ganulocytes was significantly decreased through treatment of β-CD-(D3)7/MMP-9 siRNA. In vivo fluorescence imaging assessment shows that β-CD-(D3)7/MMP-9 siRNA could not cause organ damage and organ accumulation. The results suggest that β-CD-(D3)7/MMP-9 siRNA might be developed as a novel topical agent for the diabetic wounds treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU010531 MISSION® esiRNA, esiRNA targeting human MMP9 (esiRNA1)