EMAIL THIS PAGE TO A FRIEND
Chemosphere
Effects of low levels of ultraviolet radiation on antioxidant mechanisms of Japanese Medaka (Oryzias latipes).
PMID 28448912
Abstract
Extreme weather events like drought are expected to increase with climate change, which will increase exposure of freshwater fish to ultraviolet (UV) radiation. Compared to fully grown adult fish, fish in early life stages are more susceptible to UV radiation due to the lack of well-developed pigmentation. Even though several studies have described affectation of fish health after exposure to UV radiation, most of the studies have used intensities that are only found on the surface of the earth crust or at shallow depths in water bodies, and little is known about impacts of weaker UV radiation, which can be found in deep water. This study showed effects on the antioxidant system of Japanese Medaka after 7 days of exposure to very low intensities of UV radiation, levels that can be found at deep locations in lakes and rivers. Exposure to UV radiation (UVA: 360.1 ± 18.4 μW cm
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
247502
Cumene hydroperoxide, technical grade, 80%
C9H12O2