EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Organotitania-Based Nanostructures as a Suitable Platform for the Implementation of Binary, Ternary, and Fuzzy Logic Systems.
PMID 28449282
Abstract
Titanium dioxide nanocomposites were synthesized in hierarchical architectures through the use of a 1,4-dihydroxyanthraquinone photosensitizer. In the first step, the dye was either incorporated into the TiO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q906
Quinizarin, 96%
C14H8O4