EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Ecotoxicological effects of enrofloxacin and its removal by monoculture of microalgal species and their consortium.
PMID 28449968
Abstract
Enrofloxacin (ENR), a fluoroquinolone antibiotic, has gained big scientific concern due to its ecotoxicity on aquatic microbiota. The ecotoxicity and removal of ENR by five individual microalgae species and their consortium were studied to correlate the behavior and interaction of ENR in natural systems. The individual microalgal species (Scenedesmus obliquus, Chlamydomonas mexicana, Chlorella vulgaris, Ourococcus multisporus, Micractinium resseri) and their consortium could withstand high doses of ENR (≤1 mg L
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17849
Enrofloxacin, ≥98.0% (HPLC)
C19H22FN3O3