EMAIL THIS PAGE TO A FRIEND
Food chemistry
Milk protein isolate (MPI) as a source of dipeptidyl peptidase IV (DPP-IV) inhibitory peptides.
PMID 28449998
Abstract
A multifactorial [temperature (40, 50 and 60°C), hydrolysis time (60, 150 and 240min) and enzyme to substrate ratio (E:S; 1.0, 1.5 and 2.0%)] design of experiments (DOE) was used to optimise the release of dipeptidyl peptidase IV (DPP-IV) inhibitory peptides during hydrolysis of bovine milk protein isolate (MPI) with Neutrase 0.8L™, yielding 15 hydrolysates (H1-H15). Variation in temperature and time had a significant effect on DPP-IV inhibitory properties (p<0.05) in contrast with E:S (p>0.05). The DPP-IV half maximal inhibitory concentration (IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2504
His-Leu
C12H20N4O3
I9759
Ile-Pro-Ile, ≥97% (HPLC)
C17H31N3O4
L2752
Leu-Leu
C12H24N2O3
P4126
Phe-Phe
C18H20N2O3 · xH2O