EMAIL THIS PAGE TO A FRIEND
Acupuncture in medicine : journal of the British Medical Acupuncture Society
Acupuncture upregulates G protein coupled activity in SAMP8 mice.
PMID 28450287
Abstract
Transmembrane and intracellular signal transduction of G protein is closely related to the pathophysiology of Alzheimer's disease (AD). To explore the effects of 36 senescence-accelerated (SAM) prone 8 mice were divided into three groups that remained untreated (SAMP8, n=12) or received Behavioural analysis showed that acupuncture attenuated the severe cognitive deficits observed in untreated/CA-treated SAMP8 mice. The findings of the G protein activation assays via immunoprecipitation and Western blots were that the physiologically coupled activation rate (PCAR) and maximal coupled activation rate (MCAR) of Gα