EMAIL THIS PAGE TO A FRIEND
The Journal of endocrinology
Effect of mitotane on mouse ovarian follicle development and fertility.
PMID 28450646
Abstract
Mitotane (MTT) is an adrenolytic drug used in advanced and adjuvant treatment of adrenocortical carcinoma, in Cushing's disease and in ectopic syndrome. However, knowledge about its effects on the ovary is still scarce. The purpose of this study is to investigate the effect of MTT on the ovary using