EMAIL THIS PAGE TO A FRIEND
Naunyn-Schmiedeberg's archives of pharmacology
Chronic loss of inhibitor-1 diminishes cardiac RyR2 phosphorylation despite exaggerated CaMKII activity.
PMID 28451724
Abstract
Inhibitor-1 (I-1) modulates protein phosphatase 1 (PP1) activity and thereby counteracts the phosphorylation by kinases. I-1 is downregulated and deactivated in failing hearts, but whether its role is beneficial or detrimental remains controversial, and opposing therapeutic strategies have been proposed. Overactivity of Ca
Related Materials