EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Impact of Processing on the Protein Quality of Pinto Bean (Phaseolus vulgaris) and Buckwheat (Fagopyrum esculentum Moench) Flours and Blends, As Determined by in Vitro and in Vivo Methodologies.
PMID 28452476
Abstract
Blending of protein sources can increase protein quality by compensating for limiting amino acids present in individual sources, whereas processing grain flours by extrusion or baking can also alter protein quality. To determine the effect of baking and extrusion on the protein quality of blended flours from buckwheat and pinto beans, a rodent bioassay was performed and compared to an in vitro method of protein quality determination. Overall, extruded products had higher protein efficiency ratio values, increased digestibility, and greater protein digestibility corrected amino acid score (PDCAAS) values than baked products, with the extruded buckwheat/pinto blend having the greatest PDCAAS value of the experimental diets investigated. A correlation was found between both digestibility and PDCAAS values generated from in vitro and in vivo methods. The use of in vitro digestibility analysis should be investigated as a potential replacement for the current rodent assay for nutrient content claim purposes.