EMAIL THIS PAGE TO A FRIEND
Oncology letters
Thioridazine has potent antitumor effects on lung cancer stem-like cells.
PMID 28454291
Abstract
Thioridazine (TDZ), originally an anti-psychotic drug, suppresses several types of cancer and has specificity for leukemia stem cells. The present study was performed to assess its effect on lung cancer stem-like cells, as its effect remains unknown. TDZ was utilized to treat lung cancer stem-like cells (A549 sphere cells) and its cytotoxic effect and mechanism were evaluated
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0752
Adenosine 5′-diphosphoribose sodium salt, ≥93%
C15H23N5O14P2