EMAIL THIS PAGE TO A FRIEND
Oncology letters
Overexpression of erythropoietin-producing hepatocyte receptor B4 and ephrin-B2 is associated with estrogen receptor expression in endometrial adenocarcinoma.
PMID 28454369
Abstract
Endometrial adenocarcinoma (EC) is one of the most frequently diagnosed types of endometrial cancer and is typically a consequence of continuous estrogen receptor stimulation. Erythropoietin-producing hepatocyte receptor B4 (EphB4) and its ligand ephrin-B2 have been reported to be overexpressed in EC cells; however, the function in EC remains unclear. The present study aimed to elucidate the role of EphB4 and ephrin-B2 in EC. The protein expression pattern of EphB4 and ephrin-B2 was analyzed through immunohistochemistry and western blot analysis in endometrium with adenomyosis or simple endometrial hyperplasia, atypical endometrial hyperplasia, double-positive estrogen receptor (ER)/progesterone receptor (PR) EC and double-negative ER/PR EC. The expression of EphB4 and ephrin-B2 was demonstrated to be increased in atypical EH and ER/PR-positive EC, but not ER/PR-negative EC. Furthermore, EphB4 and ephrin-B2 expression was positively associated with ER expression in EC tissue. The results of the present study suggest that the overexpression of EphB4 and ephrin-B2 in the endometrium serves a role in the pathogenesis of EC, in addition to being associated with ER expression.