EMAIL THIS PAGE TO A FRIEND
Cytotherapy
The adipose tissue-derived stromal vascular fraction cells from lipedema patients: Are they different?
PMID 28454682
Abstract
Lipedema is a hormone-related disease of women characterized by enlargement of the extremities caused by subcutaneous deposition of adipose tissue. In healthy patients application of autologous adipose tissue-derived cells has shown great potential in several clinical studies for engrafting of soft tissue reconstruction in recent decades. The majority of these studies have used the stromal vascular fraction (SVF), a heterogeneous cell population containing adipose-derived stromal/stem cells (ASC), among others. Because cell identity and regenerative properties might be affected by the health condition of patients, we characterized the SVF cells of 30 lipedema patients in comparison to 22 healthy patients. SVF cells were analyzed regarding cell yield, viability, adenosine triphosphate content, colony forming units and proliferative capacity, as well as surface marker profile and differentiation potential in vitro. Our results demonstrated a significantly enhanced SVF cell yield isolated from lipedema compared with healthy patients. In contrast, the adipogenic differentiation potential of SVF cells isolated from lipedema patients was significantly reduced compared with healthy patients. Interestingly, expression of the mesenchymal marker CD90 and the endothelial/pericytic marker CD146 was significantly enhanced when isolated from lipedema patients. The enhanced number of CD90