EMAIL THIS PAGE TO A FRIEND
Methods (San Diego, Calif.)
Identification of N
PMID 28454774
Abstract
The reversible N
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4509
4-Thiouridine, ≥98%
C9H12N2O5S