EMAIL THIS PAGE TO A FRIEND
Revista espanola de las enfermedades del aparato digestivo
[Suloctidil decreases the serotonin concentration in the brain stem of rats with a portacaval shunt].
PMID 2845525
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9384
Suloctidil, analytical standard, for drug analysis
C20H35NOS