EMAIL THIS PAGE TO A FRIEND
Recent results in cancer research. Fortschritte der Krebsforschung. Progres dans les recherches sur le cancer
Predictive assays for tumor response to single and multiple fractions of hyperthermia.
PMID 2845533