EMAIL THIS PAGE TO A FRIEND
Aging
Mitochondrial replacement in an iPSC model of Leber's hereditary optic neuropathy.
PMID 28455970
Abstract
Cybrid technology was used to replace Leber hereditary optic neuropathy (LHON) causing mitochondrial DNA (mtDNA) mutations from patient-specific fibroblasts with wildtype mtDNA, and mutation-free induced pluripotent stem cells (iPSCs) were generated subsequently. Retinal ganglion cell (RGC) differentiation demonstrates increased cell death in LHON-RGCs and can be rescued in cybrid corrected RGCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU123221 MISSION® esiRNA, esiRNA human TP53 (esiRNA1)