EMAIL THIS PAGE TO A FRIEND
Pharmacological research
Patchouli alcohol ameliorates dextran sodium sulfate-induced experimental colitis and suppresses tryptophan catabolism.
PMID 28456683
Abstract
Despite the increased morbidity of ulcerative colitis (UC) in recent years, available treatments remain unsatisfactory. Pogostemon cablin has been widely applied to treat a variety of gastrointestinal disorders in clinic for centuries, in which patchouli alcohol (PA, C