EMAIL THIS PAGE TO A FRIEND
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society
Corticotropin releasing factor receptor 1 antagonist differentially inhibits freezing behavior and changes gamma-aminobutyric acidergic activity in the amygdala in low- and high-anxiety rats.
PMID 28456768
Abstract
The aim of this study was to examine the effects of non-peptide corticotropin-releasing factor receptor 1 (CRF
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8727
Antalarmin hydrochloride, ≥98% (HPLC), solid
C24H34N4·HCl